Please login to your online account to display your discounted pricing
Corning™ PureCoat™ Amine Flat Bottom Multiwell Plates
Positively charged surface improves attachment. increases proliferation and improved differentiation for a broad range of cells that exhibit poor attachment properties in serum-free or serum-reduced conditions
$166.08 - $3,126.67
Specifications
For Use With (Application) Culture of primary cells, transfected, transformed and fastidious cell lines, culture in standard, serum-free or serum-reduced conditions, freeze-thaw recovery of cryopreserved primary cells and cell lines, including division arrested
Sterile Yes
Shelf Life 18 Months
Surface Coating Amine
View More Specs
 Alert
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Products
Catalog Number Mfr. No. Quantity Type Price Quantity    
08-772-160
corning™
C354719
5/Pk.; 5/Cs. 384-Well Amine Multiwell Microplate Case of 5 Each for $457.95
08-772-169
corning™
C356717
5/Pk.; 50/Cs. 96-Well Amine Multiwell Microplate Case of 50 Each for $2,226.08
08-772-159
corning™
C354717
5/Pk.; 5/Cs. 96-Well Amine Multiwell Microplate Case of 5 Each for $365.72
08-772-161
corning™
C354721
5/Pk.; 5/Cs. 6-Well Amine Multiwell Microplate Case of 5 Each for $183.74
08-772-162
corning™
C354723
5/Pk.; 5/Cs. 24-Well Amine Multiwell Microplate Case of 5 Each for $166.08
08-772-172
corning™
C356723
5/Pk.; 50/Cs. 24-Well Amine Multiwell Microplate Case of 50 Each for $972.87
08-772-171
corning™
C356721
5/Pk.; 50/Cs. 6-Well Amine Multiwell Microplate Case of 50 Each for $966.15
08-772-170
corning™
C356719
5/Pk.; 50/Cs. 384-Well Amine Multiwell Microplate Case of 50 Each for $3,126.67
Description & Specifications
Specifications
For Use With (Application) Culture of primary cells, transfected, transformed and fastidious cell lines, culture in standard, serum-free or serum-reduced conditions, freeze-thaw recovery of cryopreserved primary cells and cell lines, including division arrested
Sterile Yes
Shelf Life 18 Months
Surface Coating Amine
  • Chemically defined, animal-free
  • Enhances recovery from freeze-thaw
  • Suitable for primary, transfected and fastidious cell lines
  • Maintains cell monolayers during vigorous liquid handling manipulation
Safety and Handling
18 Months