Please login to your online account to display your discounted pricing

Corning™ PureCoat™ Amine Flat Bottom Multiwell Plates


Positively charged surface improves attachment. increases proliferation and improved differentiation for a broad range of cells that exhibit poor attachment properties in serum-free or serum-reduced conditions

Manufacturer: corning™  C356719

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-772-170

  • / Case of 50 Each
 

Description & Specifications

Specifications

Sterile Yes
Quantity 5/Pk.; 50/Cs.
Color Black, Clear
For Use With (Application) Culture of primary cells, transfected, transformed and fastidious cell lines, culture in standard, serum-free or serum-reduced conditions, freeze-thaw recovery of cryopreserved primary cells and cell lines, including division arrested
Shelf Life 18 Months
Surface Coating Amine
Type 384-Well Amine Multiwell Microplate
Format 384-well
Material Polystyrene
Accessories

18 Months