Please login to your online account to display your discounted pricing

Corning™ PureCoat™ Amine Flat Bottom Multiwell Plates


Positively charged surface improves attachment. increases proliferation and improved differentiation for a broad range of cells that exhibit poor attachment properties in serum-free or serum-reduced conditions

Manufacturer: corning™  C356717

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-772-169

  • / Case of 50 Each
 

Description & Specifications

Specifications

Type 96-Well Amine Multiwell Microplate
For Use With (Application) Culture of primary cells, transfected, transformed and fastidious cell lines, culture in standard, serum-free or serum-reduced conditions, freeze-thaw recovery of cryopreserved primary cells and cell lines, including division arrested
Quantity 5/Pk.; 50/Cs.
Surface Coating Amine
Shelf Life 18 Months
Material Polystyrene
Sterile Yes
Color Black, Clear
Format 96-well
Accessories

18 Months