Please login to your online account to display your discounted pricing

Corning™ PureCoat™ Amine Flasks


Positively charged surface is ideal for for preparative cell culture, cell-based, and HTS screening assays

Manufacturer: corning™  C356726

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-774-021

  • / Case of 50 Each
 

Description & Specifications

Specifications

Capacity 8.45 oz., 250mL
Closure Material Polystyrene
For Use With (Application) For preparative cell culture, cell-based and HTS screening assays
Closure Type Vented
Surface Coating Amine
Shape Rectangular
Sterile Yes
Culture Area 75cm2
Capacity (Metric) 250mL
Neck Style Straight
Material Polystyrene
Capacity (English) 8.45 oz.