Corning™ PureCoat™ Amine Culture Dishes
Positively charged surface improves attachment. increases proliferation and improved differentiation for a broad range of cells that exhibit poor attachment properties in serum-free or serum-reduced conditions
$218.85 - $537.82
Specifications
Surface Coating Amine
View More Specs
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
08-772-163 Corning™
C354732
100mm; Case of 10 Case of 10 Each for $218.85
08-772-173 Corning™
C356732
100mm; Case of 40 Case of 40 Each for $537.82
Specifications
Description
  • Chemically defined, animal-free
  • Enhance recovery from freeze-thaw
  • Suitable for primary, transfected and fastidious cell lines
  • Maintain cell monolayers during vigorous liquid handling manipulation
Description & Specifications
Specifications
Amine