Please login to your online account to display your discounted pricing
Corning™ PureCoat™ Amine Culture Dishes
Positively charged surface improves attachment. increases proliferation and improved differentiation for a broad range of cells that exhibit poor attachment properties in serum-free or serum-reduced conditions
Manufacturer: corning™  C356732
 View more versions of this product
 Alert
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Catalog No. 08-772-173
  • / Case of 40 Each
Description & Specifications
Specifications
Surface Coating Amine
Diameter (Metric) 100mm; Case of 40