Please login to your online account to display your discounted pricing

Corning™ Primaria™ Tissue Culture Dishes


Modified polystyrene surface for enhanced cell culture. Corning™ Primaria™ Tissue Culture Dishes feature flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells.

Manufacturer: corning™  C353801

 View more versions of this product

Catalog No. 08-772-4A

  • / Case of 200 Each
 

Description & Specifications

Specifications

Height (Metric) Exterior 10mm
Surface Coating Mixture of anionic and cationic functional groups
Surface Area (Metric) 11.78cm2
Sterile Yes
Diameter (Metric) 35mm
Volume (Metric) Working 2.5 to 3mL

 

  • Samples of each lot are analyzed using Electron Scanning Chemical Analysis (ESCA) to ensure a consistent ratio of nitrogen-containing functional groups
  • Suitable for growing a variety of primary cells and cell lines that grow poorly on conventional, negatively charged, standard tissue culture surfaces—including tumor cells, endothelial cells and hepatocytes
  • Features a surface treatment that is a permanent, stable modification of the polystyrene surface incorporating a mixture of anionic and cationic functional groups
  • Uniform surface chemistry created by vacuum-gas plasma treatment promotes cell attachment
  • Improved handling of small dishes with the unique Easy-Grip design
  • Nonpyrogenic
  • Sterilized by gamma irradiation
  • Packaged in peel-open, medical-style bags
  • Crystal-grade virgin polystyrene