Please login to your online account to display your discounted pricing
Corning™ Primaria™ Cell Culture Multiwell™ Plates
Features a surface treatment that is a permanent, stable modification of the polystyrene surface incorporating a mixture of anionic and cationic functional groups. Corning™ Primaria™ Cell Culture Multiwell™ Plates feature a flat bottom with low evaporation lid.
$378.49 - $514.87
Specifications
Material Polystyrene
Lid Yes
For Use With (Application) Suitable for growing a variety of primary cells and cell lines that grow poorly on conventional, negatively charged, standard tissue culture surfaces
Color Clear
Sterile Yes
View More Specs
Products
Catalog Number Mfr. No. Culture Area Format Type Volume (Metric) Well Price Quantity    
08-772-4K
corning™
C353872
0.32cm2 96-well 96-Well Cell Culture Multiwell Microplate 0.37mL Case of 50 Each for $477.46
08-772-4G
corning™
C353847
2cm2 24-well 24-Well Cell Culture Multiwell Microplate 3.5mL Case of 50 Each for $378.49
08-772-4J
corning™
C353846
9.6cm2 6-well 6-Well Cell culture Multiwell Microplate 15.5mL Case of 50 Each for $514.87
Description & Specifications
Specifications
Material Polystyrene
Lid Yes
For Use With (Application) Suitable for growing a variety of primary cells and cell lines that grow poorly on conventional, negatively charged, standard tissue culture surfaces
Color Clear
Sterile Yes
Surface Coating Corning™ Primaria
Volume (Metric) Working 1.5 to 2.2mL
Well Design Round
  • Samples of each lot are analyzed using Electron Scanning Chemical Analysis (ESCA) to ensure a consistent ratio of nitrogen-containing functional groups
  • Suitable for growing a variety of primary cells and cell lines that grow poorly on conventional, negatively charged, standard tissue culture surfaces—including tumor cells, endothelial cells and hepatocytes
  • Sterile, nonpyrogenic
  • Crystal-grade virgin polystyrene 
Individually wrapped; 50 plates per case.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.