Please login to your online account to display your discounted pricing
Corning™ Polystyrene Poly-D-Lysine Coated Microplates
Promote cell attachment
Manufacturer: corning™  C3845
 View more versions of this product
Catalog No. 08-757-468
  • / Case of 100 Each
Description & Specifications
Specifications
Lid Yes
Format 384-well
Volume (Metric) Well 95μL
Closure Yes
Material Polystyrene
For Use With (Application) Easy transition into existing automation and readers, reduced evaporation of well contents, enhanced cell attachment, reduced crosstalk and have low background fluorescence and minimal light scatter
Volume (Metric) Working 20 to 80μL
Well Design Square
Sterile Yes
Well Color Clear
Culture Area 0.06cm2
Type 384-Well Poly-D-Lysine Microplate
Surface Coating Poly-D-Lysine
Color Black, Clear