Corning™ Polystyrene Poly-D-Lysine Coated Microplates
Promote cell attachment
$2,769.69 - $6,981.27
Specifications
Material Polystyrene
Lid Yes
Sterile Yes
Surface Coating Poly-D-Lysine
Well Color Clear
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Culture Area Color Format Well Design Price Quantity  
Catalog Number Mfr. No. Culture Area Color Format Well Design Price Quantity  
08-757-464 Corning™
C3841
0.32cm2 Clear 96-well Round Case of 100 Each for $2,769.69
08-757-465 Corning™
C3842
0.32cm2 Black, Clear 96-well Round Case of 100 Each for $4,621.33
08-757-467 Corning™
C3844
0.06cm2 Clear 384-well Square Case of 100 Each for $4,688.92
08-757-468 Corning™
C3845
0.06cm2 Black, Clear 384-well Square Case of 100 Each for $6,635.78
08-757-466 Corning™
C3843
0.32cm2 White, Clear 96-well Round Case of 100 Each for $4,621.33
08-757-469 Corning™
C3846
0.06cm2 White, Clear 384-well Square Case of 100 Each for $6,981.27
Specifications
Description
  • Flat-bottom wells with lids
  • Suitable for use with both top- and bottom-reading instruments
  • Alphanumeric grid
  • Sterile and certified nonpyrogenic
  • Bulk packed
  • 96-well plates have nonreversible lids with condensation rings to reduce contamination
  • Black/Clear Bottom and White/Clear Bottom Plate Features
    • Bottoms are 60% thinner than conventional polystyrene plates, resulting in lower background fluorescence and enabling readings down to 340nm
    • Optically clear flat well bottom permits direct microscopic viewing
    • Black-walled plates have low background fluorescence, minimal light scatter, and reduced crosstalk
    • White-walled plates enhance luminescent signals and have low background luminescence and fluorescence
    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
    Description & Specifications
    Specifications
    Polystyrene
    Yes
    Clear
    Yes
    Poly-D-Lysine