Corning™ Polystyrene Disposable Culture Tubes
Manufactured from optically clear polystyrene for optimum clarity. Corning™ Polystyrene Disposable Culture Tubes are provided with threaded plug seal caps to prevent leakage.
Manufacturer:  Corning™ C430157
 View more versions of this product
Catalog No. 14-959-14
Description
Description
  • Available TC-treated or untreated
    • Tissue culture-treated tubes are available in polyfoam racks of 50 tube
    • Untreated tubes are bulk packed 25 per pack
  • Reference mark on tubes helps position cap
  • Sterilized by gamma radiation and certified nonpyrogenic
  • O.D. x L: 16 x 125mm
Specifications
Specifications
Sterile
16mm
125mm
15mL
25/Pk., 20 Pks./Cs.
15.0mL
Plug Seal Screw Cap
Polystyrene
Plug-seal screw cap
Untreated Culture Tube