Please login to your online account to display your discounted pricing
Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates permit top or bottom analysis using detection microplate readers.
Manufacturer: corning™  C3706
 View more versions of this product
Catalog No. 07-200-649
  • / Case of 100 Each
Description & Specifications
Specifications
Type EIA/RIA
Bottom Flat bottom
Binding Property Medium bind
For Use With (Application) Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
Lid No
Volume (Metric) Well 112μL
Material Polystyrene
Volume (Metric) Working 20 to 80μL
Color White
Closure No
Sterile No
  • Molded polystyrene with opaque walls and optically clear bottoms
  • Readable from top or bottom for direct microscope viewing and CCD camera-based detectors
  • Opaque black or white walls prevent well to well crosstalk
  • Recommended working volumes of 20 to 80μL; total well volume of 112μL
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Suited for flourescent and luminescent assays
  • Plate dimensions meet proposed industry standard