Please login to your online account to display your discounted pricing
Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates permit top or bottom analysis using detection microplate readers.
$2,420.82 - $2,764.62
Specifications
Lid Yes
For Use With (Application) Attachment and growth of anchorage-dependent cells
Material Polystyrene
Sterile Yes
Type 384-Well TC-treated Microplate
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Binding Property Color Sterile Price Quantity    
07-200-649
corning™
C3706
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions. Medium bind White No Case of 100 Each for $2,420.82
07-200-650
corning™
C3707
Attachment and growth of anchorage-dependent cells TC-treated White Yes Case of 100 Each for $2,764.62
Description & Specifications
Specifications
Lid Yes
For Use With (Application) Attachment and growth of anchorage-dependent cells
Material Polystyrene
Sterile Yes
Type 384-Well TC-treated Microplate
Volume (Metric) Well 112μL
Volume (Metric) Working 20 to 80μL
  • Molded polystyrene with opaque walls and optically clear bottoms
  • Readable from top or bottom for direct microscope viewing and CCD camera-based detectors
  • Opaque black or white walls prevent well to well crosstalk
  • Recommended working volumes of 20 to 80μL; total well volume of 112μL
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Suited for flourescent and luminescent assays
  • Plate dimensions meet proposed industry standard
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.