missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Polypropylene Plates
Manufactured from polypropylene for optimum visibility and high chemical resistance. Corning™ Polypropylene Plates are an excellent choice for transport and long-term storage.
Manufacturer: Corning™ C3365
Description
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or nonsterile
Specifications
Standard | |
Round | |
Clear | |
Polypropylene | |
100/Cs. | |
Non-sterile | |
96 |
Without Lid | |
U | |
Transport and longterm storage | |
100/Cs. | |
25/Pk. | |
Non-treated | |
8 x 12 |