missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Polyetherimide Centrifuge Tube Cushions

Tube cushion provides necessary support during centrifugation preventing large capacity tubes from collapsing

Manufacturer:  Corning™ C430236

 View more versions of this product

Catalog No. 05-538-53A



Description

Description

  • For use with Corning 250mL centrifuge tubes
  • Conical bottom
  • Polyetherimide
  • Gray
  • Non-sterile
  • Reusable

Specifications

Specifications

Non-sterile
8.45 oz.
Polyetherimide
1
Centrifuge Tube Cushion
Gray
250mL
Corning 250mL centrifuge tubes
Conical
Documents