missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Polyetherimide Centrifuge Tube Cushions

Tube cushion provides necessary support during centrifugation preventing large capacity tubes from collapsing

Manufacturer:  Corning™ C431124

 View more versions of this product

Catalog No. 07-200-622



Description

Description

Corning™ polyetherimide centrifuge tube cushions provide the necessary support during centrifugation to prevent these large capacity tubes from collapsing. Please check our Technical Information Listing for information on the physical and chemical properties of Corning plastic centrifuge tubes as well as important suggestions for safe use.
Specifications

Specifications

Conical
Centrifuge Tube
16.9 oz.
PP (Polypropylene)
6/Cs.
Non-sterile
Gray
500 mL
Corning 500 mL centrifuge tubes
1/Pk.
SDS
Documents