missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ PET Storage Bottles, Square with Cap
Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage
Manufacturer: Corning™ C431530
Description
Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage. PET resin is lightweight, break-resistant and transparent as glass. Bottles use a linerless, one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.
Features:
- Good for storage of media, buffers and other aqueous solutions
- Wide neck provides easier access for liquid transfer
- Screened white enamel graduations are easier to read than molded graduations
- Sterilized to SAL 10−6
- Large white marking spot for easier identification and easy opening linear tear packaging
- Individually bagged
- Validated USP Class VI, noncytotoxic and nonpyrogenic
Specifications
ISO 9001:2008 | |
GL45 | |
52.5 mm | |
Yes | |
106.5 mm | |
4.2 oz., 125 mL | |
4.2 oz. | |
2 in. | |
37 mm | |
PET | |
12/Pk. | |
Sterile | |
50 to 125 mL |
HDPE | |
Plug Seal | |
Yes | |
4.19 in. | |
Square | |
125 mL | |
Orange | |
1.4 in. | |
Easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage | |
24/Cs. | |
12 Pk. | |
Orange | |
GL45 |