Please login to your online account to display your discounted pricing

Corning™ 2.5S Nerve Growth Factor (NGF), Mouse Natural


Designed specifically for use in basic cell biology research

Manufacturer: corning™  C354005

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. CB-40005

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Formulation Lyophilized from phosphate buffered saline, carrier free, reconstitute in serum containing medium or aqueous buffer containing BSA 2%
Lyophilized Yes
Stability 3 Months
Source Mouse Submaxillary Glands
Type Nerve Growth Factor
For Use With (Application) Useful for the maintenance and differentiation of sympathetic and sensory neurons, as well as neuronal cells in-vitro
Molecular Weight 26
Purity >90% by 4 to 20% Tris Glycial Gel Electrophoresis
Quantity 10μg
Storage Requirements 2° to 8°C