Please login to your online account to display your discounted pricing

Corning™ Interleukin-2 (IL-2), Mouse Recombinant


Elicit a range of responses from many cell types

Manufacturer: corning™  C356078

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In early January 2014, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. CB-40070A

  • / Each
 

Description & Specifications

Specifications

Formulation Frozen in Dulbecco's PBS containing 0.1% BSA
Sterile Yes
Storage Requirements -70°C
Source Recombinant murine T-cell line
Shelf Life 3 Months
Purity >95% by SDS-PAGE analysis
Product Type IL-2, mouse recombinant
Quantity 10000U
Accessories

3 Months