Please login to your online account to display your discounted pricing

Corning™ Interleukin-2 (IL-2), Mouse Recombinant

Elicit a range of responses from many cell types

$363.14 - $845.35

Specifications

Formulation Frozen in Dulbecco's PBS containing 0.1% BSA
Purity >95% by SDS-PAGE analysis
Shelf Life 3 Months
Sterile Yes
Storage Requirements -70°C
View More Specs

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In early January 2014, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Products
Catalog Number Mfr. No. Quantity Product Type Price Quantity    

CB-40070

 
corning™
C354078
25000U IL-2, mouse recombinant Each for $845.35

CB-40070A

 
corning™
C356078
10000U IL-2, mouse recombinant Each for $363.14
Description & Specifications

Specifications

Formulation Frozen in Dulbecco's PBS containing 0.1% BSA
Purity >95% by SDS-PAGE analysis
Shelf Life 3 Months
Sterile Yes
Storage Requirements -70°C

Highly purified natural and recombinant biochemical.

  • Supplied frozen in buffer
  • Tested for biological activity in appropriate assays, such as the ability of growth factor to stimulate proliferation or DNA synthesis
  • Sterile filtered using a 0.2μm membrane
  • Tested for the presence of bacteria, fungi and mycoplasma

1Biological Response Modifiers Program Jurkat IL-2 reference reagent.

Safety and Handling

3 Months