Corning™ Sterile Microspatulas
Constructed of rigid nontoxic polystyrene to provide good chemical resistance to aqueous solutions. Corning™ Sterile Microspatulas are an ideal solution for molecular biology applications, lyophilized samples, and contamination-free sampling.
$329.51 - $329.51
Specifications
Certifications/Compliance USP Class VI, non-pyrogenic, RNase and DNase-free Certified
Product Type Spatula, Micro
Disposable Yes
For Use With (Application) Molecular biology applications, lyophilized samples and contamination-free sampling
Length (Metric) 15.2cm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
14-245-102 Corning™
C3013
V-shaped scoop at one end and a flat rounded blade at the other end Case of 50 Each for $329.51
14-245-100 Corning™
C3012
V-shaped scoop at one end and a flat tapered blade at the other end Case of 50 Each for $329.51
Specifications
Description
  • Single-use and disposable
  • V-shaped scoop on one end
  • RNase-/DNase-free
  • Antistatic, nonpyrogenic
  • Radation sterilized, individually packaged
  • Meets USP Class VI Requirements
Order Info
Individual packages have been radiation sterilized
Compliance
  • Nonpyrogenic certification
  • Meet USP Class VI Requirements
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
USP Class VI, non-pyrogenic, RNase and DNase-free Certified
Yes
15.2cm
Yes
Spatula, Micro
Molecular biology applications, lyophilized samples and contamination-free sampling
Polystyrene