Please login to your online account to display your discounted pricing

Microarray Storage Pouches


Protect printed microarrays from light, humidity, and environmental contaminants

Manufacturer: corning incorporated  C40086

 View more versions of this product

Catalog No. 07-202-528

  • / Case of 50 Each
 

Description & Specifications

Specifications

Product Type Storage Pouch
Color Gray
Applications Printed microarrays
Quantity 50/Cs.