Please login to your online account to display your discounted pricing

Corning™ Microarray Storage Pouches


Protect printed microarrays from light, humidity, and environmental contaminants

Manufacturer: corning™  40085

 View more versions of this product

Catalog No. 07-202-527

  • / Case of 50 Each
 

Description & Specifications

Specifications

Applications Printed microarrays
Quantity 50/Cs.
Color Gray
Product Type Storage Pouch

  • Tear-resistant, foil laminated pouches with 3 x 4 in. white marking label
  • Heat-sealable
  • Nonsterile