Please login to your online account to display your discounted pricing

Corning™ Microarray Storage Pouches

Protect printed microarrays from light, humidity, and environmental contaminants

$1,036.67 - $1,036.67

Specifications

Applications Printed microarrays
Color Gray
Quantity 50/Cs.
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

07-202-528

 
corning incorporated
C40086
Case of 50 Each for $1,036.67
Description & Specifications

Specifications

Applications Printed microarrays
Color Gray
Quantity 50/Cs.

  • Tear-resistant, foil laminated pouches with 3 x 4 in. white marking label
  • Heat-sealable
  • Nonsterile