Please login to your online account to display your discounted pricing

Corning™ Matrigel™ Membrane Matrix HC


High concentration (HC) formulation is ideal for applications in which a high protein concentration augments the growth of tumors; allows a Matrigel basement membrane matrix plug to maintain its integrity after subcutaneous injection into mice

Manufacturer: corning™  C354262

 View more versions of this product

 Alert

Note: Avoid multiple freeze-thaws; do not store in a frost-free freezer

Catalog No. 08-774-391

  • / Each
 

Description & Specifications

Specifications

Quantity 10mL
Sterile Yes
Source Engelbreth-Holm-Swarm Mouse Tumor
Storage Requirements -20°C
Without Additives Phenol Red
Format Phenol Red-Free; LDEV-Free
Product Type Basement membrane matrix
Source Engelbreth-Holm-Swarm Mouse Tumor
Quantity 10mL
Sterile Yes