Corning™ LSE Digital Dry Bath Heater Blocks
$323.16 - $759.52
Specifications
For Use With (Equipment) LSE Digital Dry Bath Heater
View More Specs
Products 4
Catalog Number Mfr. No. Type For Use With (Application) Price Quantity  
Catalog Number Mfr. No. Type For Use With (Application) Price Quantity  
07-201-781 View Documents Corning™
C480126
Dry Bath Block (Single) Holds 12 x 17mm tubes Case for $323.16
07-201-782 View Documents Corning™
C480127
Dry Bath Block (Single) Holds 6 x 20mm tubes Case for $333.26
07-201-788 View Documents Corning™
C480133
Tube Block (Dual) 384-well PCR plate Case for $759.52
07-201-786 View Documents Corning™
C480131
Tube Block (Dual) 96-well plate or 4 slides Case for $590.83
Description
Description
A variety of blocks are available for different sample containers.
Specifications
Specifications
LSE Digital Dry Bath Heater