missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Low-Volume 384-Well Polystyrene Plates
High-base design is compatible with automation and bar coding
Manufacturer: Corning™ C3540
Description
- Validated for the FLIPR™ TETRA system—outer wells aligned for proper viewing with the FLIPR system
- 2.8mm distance between well bottoms and plate bottom
- Round wells with flat bottoms
- Working volume: 5–45μL
- Untreated surface binds biomolecules through passive interactions; Nonbinding surface (NBS™) creates a nonionic hydrophilic surface that minimizes molecular interactions; Tissue culture-treated surface provides optimal cell attachment
Specifications
Flat | |
For miniaturized 384-well fluorescent homogeneous assays | |
50/Cs. | |
Non-treated | |
16 x 24 | |
0.005 to 0.04 mL |
Black, Clear Bottom | |
Polystyrene | |
Non-sterile | |
384 | |
Without Lid | |
Standard |