Corning™ Low Profile Cell Culture Flask
Unique design reduces stacking height
$721.53 - $1,269.95
Specifications
Closure Color Orange
Closure Material HDPE
Closure Type Vented
Culture Area 100cm2
Material PE (Polyethylene)
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Surface Coating Volume (Metric) Working Price Quantity  
Catalog Number Mfr. No. Surface Coating Volume (Metric) Working Price Quantity  
07-200-878 Corning™
C3816
Tissue culture treated 210mL Case of 60 Each for $721.53
13-700-433 Corning™
C3073
Corning™ Cellbind™ 30mL Case of 60 Each for $1,269.95
Specifications
Description
  • 100cm2 available growth area
  • Rectangular with canted neck, corner cap for easy pouring and improved access to flask for pipetting or scraping
  • Manufactured from optically clear virgin polystyrene
  • Available with patented Corning CellBIND™ surface treatment for enhanced cell attachment or tissue culture treatment
  • Vented cap contains 0.2μm nonwettable membrane sealed to the cap, providing consistent, sterile gas exchange while minimizing risk of contamination
  • Graduated
  • 100% integrity tested
  • Sterilized by gamma radiation and certified nonpyrogenic
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Orange
Vented
PE (Polyethylene)
For optimal cell attachment and growth
Rectangular
HDPE
100cm2
Canted
Yes
Sterile