Please login to your online account to display your discounted pricing

Corning™ Laminin, Mouse

Promotes cell adhesion, migration, growth, and differentiation, including neurite outgrowth

$231.31 - $256.96

Specifications

Concentration 1 to 10μg/cm2
Cross Reactivity Mouse
Format Liquid
Formulation Frozen in 0.05M Tris HCl, 0.15M NaCl pH 7.4
For Use With (Application) Promotes Cell Adhesion, Growth and Differentiation, Migration, Including Neurite Outgrowth
View More Specs

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Products
Catalog Number Mfr. No. Name Quantity Price Quantity    

CB-40239

 
corning™
C354239
Mouse Ultrapure Laminin 1mg Each for $256.96

CB-40232

 
corning™
C354232
Mouse Laminin 1mg Each for $231.31
Description & Specifications

Specifications

Concentration 1 to 10μg/cm2
Cross Reactivity Mouse
Format Liquid
Formulation Frozen in 0.05M Tris HCl, 0.15M NaCl pH 7.4
For Use With (Application) Promotes Cell Adhesion, Growth and Differentiation, Migration, Including Neurite Outgrowth
Molecular Weight 900kD (unreduced)
pH Range 7.4
Quality Control Testing >90% by SDS-PAGE
Source Engelbreth Holm Swarm (EHS) Mouse tumor
Storage Requirements -70°C

  • Used as thin coating on tissue-culture surfaces or as soluble additive to culture medium.
  • Major component of basement membranes
  • Tested for ability to initiate neurite outgrowth of NG-108 human neuroblastoma cells
  • Found negative for bacteria, fungi, and mycoplasma
  • Source: Engelbreth-Holm-Swarm (EHS) mouse tumor
  • Purity: >90% by SDS-PAGE (contains entactin)