Corning™ Laminin, Mouse
Promotes cell adhesion, migration, growth, and differentiation, including neurite outgrowth
Supplier: Corning™ 354232
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Laminin is a major component of basement membranes. It has numerous biological activities Âincluding promotion of cell adhesion, migration, chemotaxis, growth, and differentiation, including neurite outgrowth.
Specifications
1 to 10μg/ cm2 | |
Frozen in 0.05M Tris HCl, 0.15M NaCl pH 7.4 | |
Mouse Laminin | |
>90% by SDS-PAGE | |
Engelbreth Holm Swarm (EHS) Mouse tumor | |
Mouse |
Cell Adhesion, Cell Growth and Differentiation, Cell Migration, Neurite Outgrowth | |
900kD (unreduced) | |
7.4 | |
1 mg | |
-70°C | |
Liquid |