Corning™ Lambda Plus 8 and 12-Channel Pipettors
Designed for universal fit with all common brands of pipet tips
Manufacturer:  Corning™ C4080
 View more versions of this product
Catalog No. 07-764-706
Description
Description
Engineered to provide the highest levels of accuracy, reproducibility, and comfort
Available in single-, eight-and twelve-channel configurations, all Lambda Plus pipettors feature smooth plunger movement and extremely low pipetting forces. Volume can be set by turning either the thumb wheel or the plunger button, and a locking mechanism prevents accidental change. The Lambda Plus pipettors have been designed to maintain their calibration, even after repeated use and autoclaving. A key is included with each pipettor for easy in-house calibration.
Features
  • Factory calibrated to strict ISO 8655 standards-3-year warranty
  • Fully autoclavable and UV resistant
  • Designed for universal fit with all common brands of pipet tips
  • Adjustable tip ejector
Order Info
Accessory stand and rack available separately (Cat. Nos. 07-201-790 and -789).
Warranty
3-Year
Specifications
Specifications
±8 to ±2%
CE
Yes
8
Autoclavable
Non-sterile
3 Years
1 to 10 μL
SDS
Documents
Accessories