Corning™ Filtered IsoTip™ Universal Fit Racked Pipet Tips
Ideal for applications where avoiding cross-contamination is critical, such as DNA amplification and radioisotope handling
Supplier: Corning™ 4809
Description
- IsoTip filtered pipet tips feature an inert, hydrophobic barrier that prevents aerosolized contaminants from coming in contact with pipettor shafts. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website.)
- Ideal for applications where avoiding cross contamination is critical, such as DNA amplification and radioisotope handling.
- Packaged sterile.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifications
100 to 1000 μL | |
Sterile | |
Rack | |
Rack | |
Yes | |
Polypropylene |
Filtered | |
Natural | |
Round | |
Universal Fit | |
Isotip | |
250 μL, 500 μL, 1000 μL |