Corning™ Hybridization Chamber
Designed to hold 25 x 75mm microarray slides at a constant humidity during hybridization incubations
Manufacturer:  Corning™ C2551
 View more versions of this product
Catalog No. CS002551
Specifications Description & Specifications
Specifications
Hybridization Chamber
Wells in the base hold 10μL of water to maintain optimal interior humidity, increased interior depth allows for single slide hybridizations and for user to place two arrays face-to-face and hybridize using a single labeled target
5 Case
Polycarbonate