Corning™ Hybridization Chamber
Designed to hold 25 x 75mm microarray slides at a constant humidity during hybridization incubations
$2,582.77 - $2,582.77
Specifications
Applications Wells in the base hold 10μL of water to maintain optimal interior humidity, increased interior depth allows for single slide hybridizations and for user to place two arrays face-to-face and hybridize using a single labeled target
Material Polycarbonate
Quantity 5 Case
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
CS002551 Corning™
C2551
Hybridization Chamber Case of 5 Each for $2,582.77
Specifications
Description
  • Polycarbonate and nonsterile
  • O-ring and retaining clips assure the reusable chamber remains watertight in waterbaths and airtight in hybridization ovens
  • Wells in the base hold 10μL of water to maintain optimal interior humidity
  • Increased interior depth allows for single slide hybridizations and for user to place two arrays face-to-face and hybridize using a single labeled target
  • Used with raised-edge coverslips that are thicker or taller than standard thin coverglass
Description & Specifications
Specifications
Wells in the base hold 10μL of water to maintain optimal interior humidity, increased interior depth allows for single slide hybridizations and for user to place two arrays face-to-face and hybridize using a single labeled target
Polycarbonate
5 Case