Please login to your online account to display your discounted pricing

Corning™ Interleukin-2 (IL-2), Human Recombinant


Elicit a range of responses from many cell types

Manufacturer: corning™  C354043

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In early January 2014, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. CB-40043

  • / Each
 

Description & Specifications

Specifications

Formulation Frozen in 55mM Sodium Phosphate Buffer, containing 53mM Sucrose and 0.4% HAS
Product Type IL-2, human recombinant
Purity >96% by SDS-PAGE analysis
Shelf Life 3 Months
Storage Requirements -70°C
Reactivity Human
Sterile Yes
Source Recombinant E. coli
Quantity 10000U
Accessories

3 Months