Please login to your online account to display your discounted pricing

Corning™ GAPS™ II Coated Slides

Unique surface chemistry enables the even immobilization of DNA and uniform spot morphology

Manufacturer: corning™  40003

 View all options for this product

Catalog No. CS0040003


Description & Specifications


Coating GAPS™ II
Applications For microarray performance
Quantity 5/Pouch
Length (Metric) 25.3mm
Width (Metric) 75.5mm
Height (English) 0.04 in.
Height (Metric) 1.1mm
Sterile No
Product Type Microarray Slide

Each slide is manufactured with quality materials, then visually inspected and subjected to a rigorous cleaning process. A proprietary coating process causes the Gamma Amino Propyl Silane (GAPS) molecules to bond covalently to the glass surface, leaving positively charged primary amine groups free to attract the negatively charged sugar phosphate backbone of cDNA.

  • Ultralow fluorescent background lends superior sensitivity
  • High DNA retention provides maximum signal strength
  • To facilitate data tracking, slides are available with small bar codes traceable to manufacturing lots
  • Every lot of slides is tested to ensure superior performance
  • Dimensions: 25 × 75mm
  • Each slide is precisely packaged in a container that protects its surface during shipment