missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Rack for Conical Bottom Centrifuge Tubes

For conical bottom tubes

Manufacturer:  Corning™ C431355

 View more versions of this product

Catalog No. 07-200-611



Description

Description

These polystyrene foam racks are designed to hold 50 Corning 15 mL conical bottom plastic centrifuge tubes.
Specifications

Specifications

50
17.5 mm
Tubes (15 mL),Tubes (17.5 mm)
20/Cs.
Non-sterile
Not Autoclavable
Conical bottom centrifuge tubes
Polystyrene
20/Pk.
Centrifuge Tube Rack
SDS
Documents
Product Certifications