missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Rack for Conical Bottom Centrifuge Tubes
For conical bottom tubes
Manufacturer: Corning™ C4365
Description
These polystyrene foam racks are designed to hold conical bottom plastic centrifuge tubes.
Specifications
25 | |
Conical bottom centrifuge tubes | |
Polystyrene | |
20/Pk. | |
Centrifuge Tube Rack |
Not Autoclavable | |
Tubes (29.1 mm),Tubes (5 mL) | |
20/Cs. | |
Non-sterile |