Learn More
Corning™ FiltrEX Filter Plates
Individual filter discs eliminate liquid cross-contamination
Manufacturer: Corning™ C3504
Description
Corning FiltrEX filter microplates meet standard ANSI/SBS footprint dimensions for microplates. The rigid side walls make the microplate ideal for automation and the wide skirt accepts bar codes. Individual filter disks are encapsulated in the microplate by a unique process that ensures 100% integrity of each well. The design of the nozzle prevents sample cross-contamination and wicking. Glass fiber filter microplates can be used for a variety of applications, such as plasmid isolation, DNA purification, PCR clean-up, or receptor/ligand binding assays. They are a cost-saving alternative to DNA prep kits. Use the low-binding hydrophilic PVDF membrane for lysate clarification, protein kinase assays, or bead- or resin-based separation assays. For additional application information, visit our Technical Resources section at www.corning.com/lifesciences.
Order Info
Sterile plates are wrapped in packs of 5; 10 packs per case. Lids are not included, but may be ordered separately.
Specifications
Standard | |
Filter | |
White | |
Hydrophilic PVDF,Polystyrene | |
50/Cs. | |
50/Cs. | |
Non-sterile | |
8 x 12 |
Without Lid | |
ANSI, SBS1 | |
Ideal for automated liquid handling, centrifugation and vacuum filtration, wide skirt easily accommodates bar code labels | |
Hydrophilic PVDF | |
0.2 μm | |
10/Pk. | |
96 |