missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Fibronectin, Human

Promotes cellular attachment, spreading, and proliferation on tissue-culture surfaces. Corning™ Fibronectin, Human can be used as an additive to serum-free culture media.

Manufacturer:  Corning™ C354008

 View more versions of this product

Catalog No. CB-40008


Alert:

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.


Description

Description

 

  • Purity 90% by SDS-PAGE
  • Filtered (0.2μm membrane) and tested; found negative for the presence of bacteria, fungi, and mycoplasma
  • Lyophilized from CAPS buffer containing NaCl and CaCl2, pH 11.0

 

Specifications

Specifications

11
1mg
2° to 8°C
Fibronectin
Human Plasma
Documents