missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Epidermal Growth Factor (EGF), Mouse Natural (Receptor Grade)

Enhance the proliferation of a wide variety of cell types

Manufacturer:  Corning™ C356001

 View more versions of this product

Catalog No. CB-40001B


Alert:

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.


Description

Description

A broad range of highly purified natural and recombinant lymphokine and growth factor biochemicals designed specifically for use in basic cell biology research.

  • Mouse EGF isolated from male mouse submaxillary glands and supplied in lyophilized form
  • Receptor grade EGF has been further purified by DEAE chromatography to >98% purity
  • Human EGF obtained as a recombinant from E. coli and supplied in lyophilized form
  • Products are tested for biological activity in appropriate assays, such as the ability of growth factors to stimulate proliferation or DNA synthesis
  • Lymphokines and growth factors are also sterile filtered generally using a 0.2μm membrane
  • Tested for the presence of bacteria, fungi and mycoplasma

Specification sheets include detailed information on quality control testing, product source, storage, stability and recommendations on proper experimental concentrations.

Specifications

Specifications

Epidermal Growth Factor
A broad range of highly purified natural and recombinant lymphokine and growth factor biochemicals designed specifically for use in basic cell biology research
Lyophilized from ammonium acetate, carrier free, reconstitute in dH2O
Yes
>95% by SDS-PAGE and >80% by pH 9.9 PAGE
3 Months
Male Mouse Submaxillary Glands
Mouse
Natural
Culture
6.1
1mg (10 x 100μg)
2° to 8°C
Documents