missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Easy Grip Polystyrene Storage Bottles

Manufacturer:  Corning™ C11650

 View more versions of this product

Catalog No. 15-100-120



Description

Description

  • Double-bagged
  • Threaded
  • With dip tube
  • Male luer lock tubing
  • 0.2um MLL/FLL filter

Specifications

Specifications

5.07 oz., 150mL
5.07 oz.
1.7 in.
Yes
Polystyrene
8 Cs.
Orange
MLL/FLL
150mL
HDPE
Yes
Easy grip
8/Cs.
Sterile
45mm
Documents