Corning™ DNA-BIND™ 96-Well Plates
Convenient method for covalently immobilizing ssDNA by either the 5« or 3« end
Manufacturer:  Corning™ C2525
 View more versions of this product
Catalog No. 07-200-586
Description
Description
  • Ready to use without surface preparation, prehybridization, or addition of expensive crosslinking reagents or highly purified oligos
  • Ideal for hybridization and amplification
  • Surface provides binding with high specificity and extremely low background
  • Flat-bottom wells
Specifications
Specifications
8 x 12
Ideal for hybridization and amplification
10/Cs.
1/Pk.
Clear
Without Lid
Round
Clear
Polystyrene
10/Cs.
Non-sterile
96
Standard