Please login to your online account to display your discounted pricing

Corning™ DNA-BIND™ 96-Well Plates


Convenient method for covalently immobilizing ssDNA by either the 5« or 3« end

Manufacturer: corning™  C2499

 View more versions of this product

Catalog No. 07-200-680

  • / Case of 50 Each
 

Description & Specifications

Specifications

Well Color White
Well Design Round
Material Polystyrene
Volume (Metric) Well 360μL
Quantity 50/Cs.
Skirt Style Standard
Sterile No
Color White
For Use With (Application) Ideal for hybridization and amplification