Please login to your online account to display your discounted pricing

Corning™ Disposable Sterile Bottles


Use as replacement receivers for Corning Disposable Sterile Filter Systems or as storage bottles. Corning™ Disposable Sterile Bottles are easy to grip and ideal for storage of media, buffers and other aqueous solutions.

Manufacturer: corning™  C430282

 View more versions of this product

Catalog No. 09-761-10

  • / Case of 24 Each
 

Description & Specifications

Specifications

Quantity 24 Case
Disposable Yes
Item Description For use with 4.5cm bottle neck
Sterile Yes
Volume (Metric) 500mL
Includes Orange HDPE plug seal
Product Type Graduated Easy Grip Storage Bottle
Material Polystyrene
Description Applications: Storage of media, buffers and other aqueous solutions

 

  • Durable polystyrene bottles
  • Graduated
  • Neck size: 45mm
  • Easy-to-read graduations
  • Plug seal cap provides an airtight seal and helps minimize the risk of contamination
  • With sterile caps