Please login to your online account to display your discounted pricing

Corning™ Disposable Sterile Bottles


Use as replacement receivers for Corning Disposable Sterile Filter Systems or as storage bottles. Corning™ Disposable Sterile Bottles are easy to grip and ideal for storage of media, buffers and other aqueous solutions.

Manufacturer: corning™  C431175

 View more versions of this product

Catalog No. 09-761-140

  • / Case of 24 Each
 

Description & Specifications

Specifications

Description Applications: Storage of media, buffers and other aqueous solutions
Includes Orange HDPE plug seal
Item Description For use with 4.5cm bottle neck
Product Type Graduated Easy Grip Storage Bottle
Quantity 24 Case
Material Polystyrene
Sterile Yes
Disposable Yes
Volume (Metric) 150mL

 

  • Durable polystyrene bottles
  • Graduated
  • Neck size: 45mm
  • Easy-to-read graduations
  • Plug seal cap provides an airtight seal and helps minimize the risk of contamination
  • With sterile caps