missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Disposable Sterile Bottles

Use as replacement receivers for Corning Disposable Sterile Filter Systems or as storage bottles. Corning™ Disposable Sterile Bottles are easy to grip and ideal for storage of media, buffers and other aqueous solutions.

Manufacturer:  Corning™ C430518

 View more versions of this product

Catalog No. 09-761-11


 

Specifications

Description

Bottle, Sterile; Corning ; Disposable; Polystyrene; Neck size 45mm; Easy grip sides facilitate handling; Plug seal provides airtight seal and minimizes risk of contamination; Non-Pyrogenic; Red HDPE cap; 1000mL; 24/Cs.

 

  • Durable polystyrene bottles
  • Graduated
  • Neck size: 45mm
  • Easy-to-read graduations
  • Plug seal cap provides an airtight seal and helps minimize the risk of contamination
  • With sterile caps

 

Description & Specifications

Specifications

24 Case
For use with 4.5cm bottle neck
Yes
1000mL
Polystyrene
Yes
Applications: Storage of media, buffers and other aqueous solutions
Orange HDPE plug seal
Graduated Easy Grip Storage Bottle