Corning™ Disposable Spinner Flask Aseptic Transfer Caps
For use with Corning disposable spinner flasks with top cap and angled sidearms
Manufacturer:  Corning™ C3528
 View more versions of this product
Catalog No. 07-200-341
Description
Description
These disposable Corning™ Spinner Flask Aseptic Transfer Caps will both reduce contamination and enhance the productivity of your cell culture operations when adding or removing medium and cells. Aseptic Transfer Caps are sterile (10−6 SAL), individually double bagged and fit the side arms on 500 mL Corning™ Disposable Spinner Flasks (Cat. No. 3153).
  • Cap is made of high density polyethylene; autoclaving is not recommended.
  • The venting port ends in a 25mm diameter vent filter with a 0.2 μm pore PVDF membrane and a barbed fitting.
  • The liquid transfer port has an internal end with a rigid polypropylene dip tube angled to reach the bottom of the flask and on the external side a 1/4 inch ID (6.35mm), 1/4 inch OD chemically resistant, heat sealable flexible tubing, ending with Medical Grade Polycarbonate Quick Connect.
  • Specifications
    Specifications
    Orange
    Polycarbonate
    Sidearm 0.25 in. dip tube with 0.2μm pore, vent, medical grade
    1/Pk.
    GL 45
    54 mm
    500 mL Disposable spinner flask
    2/Cs.
    45°