missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Disposable Roller Bottles

Manufactured from optically clear virgin polystyrene for excellent visibility and tissue culture-treated for optimal cell attachment. Corning™ Disposable Roller Bottles allow Initial scale-up of adherent cells with intermittent exposure of cells to the air and the media.

Manufacturer:  Corning™ C431198

 View more versions of this product

Catalog No. 06-443-8



Description

Description

  • Bottles are manufactured from virgin polystyrene, caps from high density polyethylene
  • Both resins meet the USP Class VI requirements for plastic containers and closures
  • Treated for optimal cell attachment
  • One piece seamless construction with printed graduations
  • Printed lot numbers to aid in product traceability
  • Sterilized by gamma radiation and nonpyrogenic
Specifications

Specifications

HDPE
Vented
10.66 in.
Wide
2200 mL
1.77 in.
Polystyrene
Sterile
Red
117mm
48 mm
Yes
271 mm
74.3 oz., 2200mL
74.3 oz.
45mm
36/case
850 cm2
4.6 in.
Tissue culture treated
SDS
Documents
Product Certifications

Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.