missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Polycarbonate Erlenmeyer Flasks

Manufactured from optically clear polycarbonate for optimum visibility and durability. Corning™ Disposable Polycarbonate Erlenmeyer Flasks are ideal for shaker cultures applications.

Manufacturer:  Corning™ C431146

 View more versions of this product

Catalog No. 07-200-671



Description

Description

  • Made from optically clear polycarbonate
  • Ideal for shaker culture applications
  • Vent caps available for applications requiring sterile gas exchange
  • Sterilized by gamma radiation and certified nonpyrogenic
  • RNase- and DNase-free
Specifications

Specifications

33.8 oz.
Polypropylene
4.3 cm
Polycarbonate
50 mL
25/Cs.
Yes
Flat top
Orange
Top style: Threaded
1 L
For shaker culture applications
Wide Mouth
1/Pk.
Sterile
SDS
Documents